Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclohexenylboronic acid pinacol ester 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43031-0010 43031-5000
TS43031-0010EA 243 USD InStock
TS43031-0010 TS43031-5000
1-Cyclohexenylboronic acid pinacol ester 97%
1-Cyclohexenylboronic acid pinacol ester


Formula: C₁₂H₂₁BO₂
Boiling Pt: 125 °C (.2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD05663845
CAS Number: 141091-37-4
Order Now
Specification Test Results
Appearance Clear colorless to tan Liquid
Infrared spectrum Conforms
GC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty