Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Butylpicolinic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

19896-0010 19896-2500
TS19896-0010EA 258 USD InStock
TS19896-0010 TS19896-2500
5-Butylpicolinic acid 99%
5-Butylpicolinic acid


Formula: C₁₀H₁₃NO₂
Melting Pt: 97…101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006298
CAS Number: 536-69-6
EINECS: 208-643-0
Order Now
Specification Test Results
Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 101 °C
Titration with NaOH ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty