Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-chloro-2-(hydroxyimino)acetate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39874-0050 39874-0250
TS39874-0050EA 116 USD InStock
TS39874-0050 TS39874-0250
Ethyl-2-chloro-2-(hydroxyimino)acetate 97%
Ethyl-2-chloro-2-(hydroxyimino)acetate
Appearance: White to yellow Crystalline powder


Formula: C₄H₆ClNO₃
Melting Pt: 70…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010209
CAS Number: 14337-43-0
UN: 3335
ADR: 9,III
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 75 °C
Titration Argentometric ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty