Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-1,6-diaminohexane 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30183-0010 30183-0050
TS30183-0010EA 75 USD InStock
TS30183-0010 TS30183-0050
N-Boc-1,6-diaminohexane 95%
N-Boc-1,6-diaminohexane
Appearance: Clear light yellow Viscous liquid


Formula: C₁₁H₂₄N₂O₂
MW: 216.33 g/mol
Boiling Pt: 106…110 °C (.3 mmHg)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00671489
CAS Number: 51857-17-1
UN: 2735
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty