Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminopropylphosphonic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34494-0010 34494-0050
TS34494-0010EA 148 USD InStock
TS34494-0010 TS34494-0050
3-Aminopropylphosphonic acid 97%
3-Aminopropylphosphonic acid


Formula: C3H10NO3P
MW: 139.0902 g/mol
Melting Pt: 290…294 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008222
CAS Number: 13138-33-5
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty