Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodoxybenzoic acid stabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

37465-0100 37465-0500
TS37465-0100EA 196 USD InStock
TS37465-0100 TS37465-0500
2-Iodoxybenzoic acid stabilised
2-Iodoxybenzoic acid
Appearance: White to off-white Powder or prills and/or agglomerates


Formula: C₇H₅IO₄
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD02912492
CAS Number: 61717-82-6
UN: 1759
Order Now
Specification Test Results
Appearance White to beige Powder or crystals
Titration Iodometric 39 to 50 % as 2-Iodoxybenzoic acid
Identification HPLC (2-Iodoxybenzoic acid, 2-Iodosobenzoic acid, Isophthalic acid, Benzoic acid)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty