Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 97+% stabilized

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.05%

39553-0010 39553-0050
TS39553-0010EA 68 USD InStock
TS39553-0010 TS39553-0050
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 97+% stabilized
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane
Appearance: Clear colorless to amber Liquid


Formula: C₈H₁₅BO₂
Boiling Pt: 40 °C (20 mmHg)
Density: 0.908 g/cm³
Flash Pt: 34 °C
Storage Temperature: Freezer
MDL Number: MFCD00192492
CAS Number: 75927-49-0
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥97.0 %
Stabilizer ≤500 ppm BHT

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty