Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Diaminostilbene-2,2'-disulphonic Acid 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

40641-1000 40641-5000
TS40641-1000EA 91 USD InStock
TS40641-1000 TS40641-5000
4,4'-Diaminostilbene-2,2'-disulphonic Acid 95%
4,4'-Diaminostilbene-2,2'-disulfonic Acid
Appearance: Cream to yellow-brown Crystalline powder


Formula: C₁₄H₁₄N₂O₆S₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00024946
CAS Number: 81-11-8
Order Now
Specification Test Results
Appearance Cream to yellow-brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty