Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Acridinecarboxylic acid hydrate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

40022-0010 40022-0050 40022-0250
TS40022-0010EA 70 USD InStock
TS40022-0010 TS40022-0050 TS40022-0250
9-Acridinecarboxylic acid hydrate 97%
9-Acridinecarboxylic acid hydrate


Formula: C₁₄H₁₁NO₃
MW: 241.24 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00149578
CAS Number: 332927-03-4
Order Now
Specification Test Results
Appearance Bright yellow to yellow-green Crystalline powder
Infrared spectrum Conforms
Water ≤15 % (K.F.)
Titration with NaOH ≥96.0 % (on dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty