Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (chloromethyl)phosphonate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33065-0010 33065-0050
TS33065-0010EA 54 USD InStock
TS33065-0010 TS33065-0050
Diethyl (chloromethyl)phosphonate 98%
Diethyl (chloromethyl)phosphonate
Appearance: Clear colorless to light yellow Liquid


Formula: C₅H₁₂ClO₃P
Boiling Pt: 109…110 °C (10 mmHg)
Density: 1.2 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00010189
CAS Number: 3167-63-3
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty