Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-bromoisovalerate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

40979-0100 40979-0500
TS40979-0100EA 71 USD InStock
TS40979-0100 TS40979-0500
Ethyl-2-bromoisovalerate 95%
Ethyl-2-bromoisovalerate


Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
Boiling Pt: 77 °C (12 mmHg)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00026855
CAS Number: 609-12-1
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4510 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty