Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2-bromoethanesulfonate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16003-0250 16003-1000
TS16003-0250EA 38 USD InStock
TS16003-0250 TS16003-1000
Sodium-2-bromoethanesulfonate 98%
Sodium 2-Bromoethanesulfonate
Appearance: White to off-white Crystalline powder


Formula: C₂H₅BrO₃S
Melting Pt: 282…286 °C
Storage Temperature: Ambient
MDL Number: MFCD00007530
CAS Number: 4263-52-9
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 282 to 286 °C
Titration Argentometric ≥97.5 % (ex Br) On dry substance
Loss on drying ≤0.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty