Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2-propylvalerate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

27183-0050 27183-0250
TS27183-0050EA 28 USD InStock
TS27183-0050 TS27183-0250
Sodium-2-propylvalerate 98%
Sodium-2-propylvalerate
Appearance: White Powder


Formula: C₈H₁₅O₂Na
MW: 166.20 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00078604
CAS Number: 1069-66-5
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty