Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-bromocyclobutanecarboxylate 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18871-0010 18871-0050
TS18871-0010EA 21 USD InStock
TS18871-0010 TS18871-0050
Ethyl-1-bromocyclobutanecarboxylate 96%
Ethyl-1-bromocyclobutanecarboxylate
Appearance: Clear colorless Liquid


Formula: C₇H₁₁BrO₂
Boiling Pt: 85…88 °C (12 mmHg)
Density: 1.279 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001322
CAS Number: 35120-18-4
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20°C, 589 nm)
GC ≥95 %
HPLC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty