Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-3-methyl-2-oxobutanoate 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

18972-0010 18972-0050 18972-0250
TS18972-0010EA 78 USD InStock
TS18972-0010 TS18972-0050 TS18972-0250
Sodium-3-methyl-2-oxobutanoate 98+%
Sodium-3-methyl-2-oxobutanoate


Formula: C₅H₇NaO₃
MW: 138.098 g/mol
Melting Pt: 227…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00002581
CAS Number: 3715-29-5
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 227 to 231 °C
Titration with HClO4 ≥98.0 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty