Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl vinyl ether 98% stabilized

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

37912-0100 37912-0500
TS37912-0100EA 119 USD InStock
TS37912-0100 TS37912-0500
tert-Butyl vinyl ether 98% stabilized
tert-Butyl vinyl ether
Appearance: Clear colorless to light yellow Liquid


Formula: C₆H₁₂O
Boiling Pt: 78 °C (760 mmHg)
Density: 0.762 g/cm³
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048246
CAS Number: 926-02-3
UN: 3271
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.3990 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Approx. 1000 ppm KOH

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty