Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-Dibenzoyl-L-tartaric anhydride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

33603-0050 33603-0250
TS33603-0050EA 52 USD InStock
TS33603-0050 TS33603-0250
(+)-Dibenzoyl-L-tartaric anhydride 98%
(+)-Dibenzoyl-L-tartaric anhydride
Appearance: White to off-white Crystalline powder


Formula: C₁₈H₁₂O₇
MW: 340.287 g/mol
Melting Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00067102
CAS Number: 64339-95-3
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
Titration by coupling ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty