Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Tetrazol-5-yl)phenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44557-0010 44557-0050
TS44557-0010EA 140 USD InStock
TS44557-0010 TS44557-0050
4-(Tetrazol-5-yl)phenylboronic acid 97%
4-(Tetrazol-5-yl)phenylboronic acid


Formula: C₇H₇BN₄O₂
MW: 189.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739099
CAS Number: 179942-55-3
Order Now
Specification Test Results
Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥95.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty