Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloromethylphosphonic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

45160-0010 45160-0050
TS45160-0010EA 104 USD InStock
TS45160-0010 TS45160-0050
Chloromethylphosphonic acid 98%
Chloromethylphosphonic acid


Formula: CH₄ClO₃P
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00013955
CAS Number: 2565-58-4
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 92 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty