Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,2,2-Tetrabromoethane 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

18087-0010 18087-0025 18087-2500
TS18087-0010EA 145 USD InStock
TS18087-0010 TS18087-0025 TS18087-2500
1,1,2,2-Tetrabromoethane 98%
1,1,2,2-Tetrabromoethane


Formula: Br₂HCCHBr₂
Storage Temperature: Ambient
MDL Number: MFCD00000133
CAS Number: 79-27-6
EINECS: 201-191-5
UN: 2504
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecc5c2f2399032fc65a1f90353ec556de6f2f655&vid=ecc5c2f2399032fc65a1f90353ec556de6f2f655&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.6355 to 1.6375 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤75 APHA

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty