Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-5-chloropentane 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

22121-0050 22121-0250 22121-1000
TS22121-0050EA 27 USD InStock
TS22121-0050 TS22121-0250 TS22121-1000
1-Bromo-5-chloropentane 98%
1-Bromo-5-chloropentane


Formula: BrH₂C(CH₂)₃CH₂Cl
Boiling Pt: 210…212 °C (760 mmHg)
Density: 1.4 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00001016
CAS Number: 54512-75-3
EINECS: 259-193-7
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4826 to 1.4846 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty