Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iododecane 98% stabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33036-5000 33036-0250 33036-1000
TS33036-5000EA 487 USD InStock
TS33036-5000 TS33036-0250 TS33036-1000
1-Iododecane 98% stabilised
1-Iododecane


Formula: C₁₀H₂₁I
Boiling Pt: 124 °C (11 mmHg)
Density: 1.25 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00001085
CAS Number: 2050-77-3
EINECS: 218-104-1
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4840 to 1.4870 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer copper

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty