Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodooctane 98% stabilized

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41246-1000 41246-5000
TS41246-1000EA 95 USD InStock
TS41246-1000 TS41246-5000
1-Iodooctane 98% stabilized
1-Iodooctane


Formula: C₈H₁₇I
Boiling Pt: 225…226 °C (760 mmHg)
Melting Pt: 46…45 °C
Density: 1.33 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00001105
CAS Number: 629-27-6
EINECS: 211-084-5
Order Now
Specification Test Results
Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4868 to 1.4888 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer copper
Acidity (CH3COOH) ≤0.002 %
Peroxides ≤0.005 % (as H2O2)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty