Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodo-2-methylpropane 97% stabilized

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22374-0250 22374-1000 22374-5000
TS22374-0250EA 85 USD InStock
TS22374-0250 TS22374-1000 TS22374-5000
1-Iodo-2-methylpropane 97% stabilized
1-Iodo-2-methylpropane


Formula: C₄H₉I
Boiling Pt: 120…121 °C (760 mmHg)
Melting Pt: –93 °C
Density: 1.59 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00001084
CAS Number: 513-38-2
EINECS: 208-160-5
UN: 2391
ADR: 3,II
Merck Index: 1305160
Order Now
Specification Test Results
Appearance Clear colorless to yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer copper

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty