Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Decyne 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

22343-0050 22343-0250 22343-1000
TS22343-0050EA 29 USD InStock
TS22343-0050 TS22343-0250 TS22343-1000
1-Decyne 98%
1-Decyne


Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 174
Melting Pt: -44
Density: 0.76 g/cm³
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009576
CAS Number: 764-93-2
EINECS: 212-132-8
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty