Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromothiophenol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22198-0010 22198-0100 22198-0500
TS22198-0010EA 29 USD InStock
TS22198-0010 TS22198-0100 TS22198-0500
2-Bromothiophenol 97%
2-Bromothiophenol


Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 128
Density: 1.604 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004827
CAS Number: 6320-02-1
EINECS: 228-665-4
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6340 to 1.6370 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty