Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-3-Carene

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

46399-0050 46399-0250
TS46399-0050EA 29 USD InStock
TS46399-0050 TS46399-0250
(+)-3-Carene
(+)-3-Carene


Formula: C₁₀H₁₆
Boiling Pt: 170 °C
Density: 0.865
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00066417
CAS Number: 498-15-7
UN: 2319
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4760 (20°C, 589 nm)
Density 0.854 to 0.884 kg/l
GC ≥88 %
Specific optical rotation +13.0° to +17.0° (20°C, 589 nm) (neat)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty