Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Quinolinol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41882-0010 41882-0050
TS41882-0010EA 189 USD InStock
TS41882-0010 TS41882-0050
7-Quinolinol 99%
7-Quinolinol


Formula: C₉H₇NO
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 239…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00016730
CAS Number: 580-20-1
EINECS: 209-457-2
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 239 to 244 °C (Dec)
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty