Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Bromomethyl)cyclobutane 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36764-0050 36764-0250 36764-1000
TS36764-0050EA 110 USD InStock
TS36764-0050 TS36764-0250 TS36764-1000
(Bromomethyl)cyclobutane 99%
(Bromomethyl)cyclobutane


Formula: C₅H₉Br
Boiling Pt: 145…148 °C (760 mmHg)
Density: 1.339 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00040960
CAS Number: 17247-58-4
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4830 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty