Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylcyclohexanone 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

34619-0050 34619-0250
TS34619-0050EA 40 USD InStock
TS34619-0050 TS34619-0250
2-Acetylcyclohexanone 97%
2-Acetylcyclohexanone


Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Density: 1.063 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001633
CAS Number: 874-23-7
EINECS: 212-858-5
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5070 to 1.5130 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty