Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzoyl fluoride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40204-0050 40204-0250
TS40204-0050EA 39 USD InStock
TS40204-0050 TS40204-0250
Benzoyl fluoride 97%
Benzoyl fluoride


Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 159
Melting Pt: -28.5
Density: 1.15 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000364
CAS Number: 455-32-3
UN: 2920
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty