Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-4-quinolinecarboxylic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44993-0010 44993-0050
TS44993-0010EA 134 USD InStock
TS44993-0010 TS44993-0050
3-Hydroxy-4-quinolinecarboxylic acid 97%
3-Hydroxy-4-quinolinecarboxylic acid


Formula: C₁₀H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD01536959
CAS Number: 118-13-8
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty