Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl isothiocyanate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40213-0050 40213-0250 40213-1000
TS40213-0050EA 34 USD InStock
TS40213-0050 TS40213-0250 TS40213-1000
Benzyl isothiocyanate 98%
Benzyl isothiocyanate


Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 243
Melting Pt: 24
Density: 1.12 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004819
CAS Number: 622-78-6
EINECS: 210-753-9
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6010 to 1.6040 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty