Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-3-Butyne-2-ol 95%, 98% ee

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

36241-0010 36241-0050
TS36241-0010EA 119 USD InStock
TS36241-0010 TS36241-0050
(S)-(-)-3-Butyne-2-ol 95%, 98% ee
(S)-(-)-3-Butyne-2-ol


Formula: C₄H₆O
MDL Number: MFCD00190166
CAS Number: 2914-69-4
UN: 2929
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty