Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoroindole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34251-0010 34251-0050 34251-2500
TS34251-0010EA 52 USD InStock
TS34251-0010 TS34251-0050 TS34251-2500
6-Fluoroindole 98%
6-Fluoroindole


Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9
Order Now
Specification Test Results
Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 77 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty