Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl disulphide 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16559-0010 16559-0025 16559-0500 16559-2500
TS16559-0010EA 89 USD InStock
TS16559-0010 TS16559-0025 TS16559-0500 TS16559-2500
Dimethyl disulphide 99%
Dimethyl disulphide


Formula: C₂H₆S₂
MW: 94.2 g/mol
Boiling Pt: 110
Melting Pt: -98
Density: 1.0625 g/cm³
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008561
CAS Number: 624-92-0
EINECS: 210-871-0
UN: 2381
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5243 to 1.5263 (20°C, 589 nm)
Water ≤0.06 %
GC ≤1 % (Methylmercaptan content)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty