Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl trisulphide 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41503-0050 41503-0250
TS41503-0050EA 75 USD InStock
TS41503-0050 TS41503-0250
Dimethyl trisulphide 98+%
Dimethyl trisulphide


Formula: C₂H₆S₃
MW: 126.27
Melting Pt: –68 °C
Flash Pt: 52 °C
MDL Number: MFCD00039808
CAS Number: 3658-80-8
EINECS: 222-910-9
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5995 to 1.6025 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty