Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butanesulphonyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40332-0250 40332-1000
TS40332-0250EA 95 USD InStock
TS40332-0250 TS40332-1000
1-Butanesulphonyl chloride 98%
1-Butanesulphonyl chloride


Formula: C₄H₉ClO₂S
MW: 156.63 g/mol
Boiling Pt: 208
Melting Pt: -25
Density: 1.2 g/cm³
Flash Pt: 79 °C
MDL Number: MFCD00007464
CAS Number: 2386-60-9
EINECS: 219-204-8
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4570 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty