Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Anisoyl chloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21297-0050 21297-0250 21297-1000
TS21297-0050EA 39 USD InStock
TS21297-0050 TS21297-0250 TS21297-1000
3-Anisoyl chloride 99%
3-Anisoyl chloride


Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 123
Density: 1.214 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00000673
CAS Number: 1711-05-3
EINECS: 216-975-2
UN: 1729
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5595 (20°C, 589 nm)
GC ≥98.5 %
Titration RCOCl ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty