Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Phenylene diisothiocyanate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

41751-0050 41751-0250
TS41751-0050EA 107 USD InStock
TS41751-0050 TS41751-0250
1,4-Phenylene diisothiocyanate 99%
1,4-Phenylene diisothiocyanate


Formula: C₈H₄N₂S₂
MW: 192.27 g/mol
Boiling Pt: 319
Melting Pt: 131
Storage Temperature: Refrigerator
MDL Number: MFCD00004811
CAS Number: 4044-65-9
EINECS: 223-741-3
UN: 2811
ADR: 6.1 III
Merck Index: 13,01304
Order Now
Specification Test Results
Appearance Off-white to yellow to beige to light grey Crystalline powder or needles
Infrared spectrum Conforms
Melting point 129 to 133 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty