Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethynylanisole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

37992-0010 37992-0050
TS37992-0010EA 77 USD InStock
TS37992-0010 TS37992-0050
4-Ethynylanisole 97%
4-Ethynylanisole


Formula: C₉H₈O
Boiling Pt: 87…91 °C (11 mmHg)
Melting Pt: 28…30 °C
Density: 1.019 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00168815
CAS Number: 768-60-5
Order Now
Specification Test Results
Appearance Clear colorless to yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5625 to 1.5675 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty