Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,6-difluoroaniline 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35020-0010 35020-0050
TS35020-0010EA 97 USD InStock
TS35020-0010 TS35020-0050
2-Chloro-4,6-difluoroaniline 97%
2-Chloro-4,6-difluoroaniline


Formula: C₆H₄ClF₂N
Melting Pt: 28…33 °C
MDL Number: MFCD00142892
CAS Number: 36556-56-6
Order Now
Specification Test Results
Appearance Off-white to grey-green Low melting solid
Infrared spectrum Conforms
Melting point 28 to 33 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty