Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Hexyl-3-methylimidazolium hexafluorophosphate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35418-0050 35418-0250 35418-1000
TS35418-0050EA 76 USD InStock
TS35418-0050 TS35418-0250 TS35418-1000
1-Hexyl-3-methylimidazolium hexafluorophosphate 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate


Formula: C₁₀H₁₉F₆N₂P
Melting Pt: –73.5 °C
Density: 1.3045 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD03093296
CAS Number: 304680-35-1
Order Now
Specification Test Results
Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4200 (20°C, 589 nm)
NMR ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty