Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorothiazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45726-0010 45726-0050 45726-0250
TS45726-0010EA 40 USD InStock
TS45726-0010 TS45726-0050 TS45726-0250
2-Chlorothiazole 97%
2-Chlorothiazole


Formula: C₃H₂ClNS
Boiling Pt: 145…146 °C (760 mmHg)
Density: 1.378 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00210701
CAS Number: 3034-52-4
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty