Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1-hexyne 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38081-0050 38081-0250
TS38081-0050EA 65 USD InStock
TS38081-0050 TS38081-0250
6-Chloro-1-hexyne 97%
6-Chloro-1-hexyne


Formula: C₆H₉Cl
Flash Pt: 37 °C
MDL Number: MFCD00013697
CAS Number: 10297-06-0
UN: 1993
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty