Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromoisobutyric acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30520-0050 30520-1000 30520-5000
TS30520-0050EA 15 USD InStock
TS30520-0050 TS30520-1000 TS30520-5000
2-Bromoisobutyric acid 98%
2-Bromoisobutyric acid


Formula: C₄H₇BrO₂
Boiling Pt: 198…200 °C (760 mmHg)
Melting Pt: 41…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004168
CAS Number: 2052-01-9
EINECS: 218-139-2
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to yellow to orange Semitransparent crystalline powder or crystals or mass and/or chunks
Infrared spectrum Conforms
Melting point 41 to 46 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty