Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Patent blue V sodium salt

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

33933-0050 33933-0250 33933-1000
TS33933-0050EA 24 USD InStock
TS33933-0050 TS33933-0250 TS33933-1000
Patent blue V sodium salt
Patent blue V sodium salt


Formula: C₂₇H₃₁N₂NaO₇S₂
Storage Temperature: Ambient
MDL Number: MFCD09608138
CAS Number: 20262-76-4
EINECS: 222-573-8
Order Now
Specification Test Results
Appearance Dark blue-green Crystalline powder
Infrared spectrum Conforms
Dye content ≥80 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty