Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroethanesulfonyl chloride 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40461-0050 40461-0250
TS40461-0050EA 42 USD InStock
TS40461-0050 TS40461-0250
2-Chloroethanesulfonyl chloride 95%
2-Chloroethanesulphonyl chloride


Formula: C₂H₄Cl₂O₂S
Boiling Pt: 78 °C (9 mmHg)
Melting Pt: 3 °C
Density: 1.55 g/cm³
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00007461
CAS Number: 1622-32-8
EINECS: 216-594-1
UN: 2927
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
Titration Argentometric ≥94 % (ex Cl)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty