Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propargyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

45184-0050 45184-0250
TS45184-0050EA 40 USD InStock
TS45184-0050 TS45184-0250
Propargyl chloride 98%
Propargyl chloride


Formula: C₃H₃Cl
Boiling Pt: 58…61 °C (760 mmHg)
Melting Pt: –77 °C
Density: 1.03 g/cm³
Flash Pt: 19 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000980
CAS Number: 624-65-7
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4370 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty