Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-3-buten-1-ol 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36821-0100 36821-0500
TS36821-0100EA 231 USD InStock
TS36821-0100 TS36821-0500
3-Bromo-3-buten-1-ol 97+%
3-Bromo-3-buten-1-ol


Formula: C₄H₇BrO
Boiling Pt: 175 °C (760 mmHg)
Density: 1.522 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00154041
CAS Number: 76334-36-6
Order Now
Specification Test Results
Appearance Clear brown Liquid
Infrared spectrum Conforms
Refractive index 1.5000 to 1.5020 (20°C, 589 nm)
GC ≥97.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty